Anti-KLRB1C Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 694370]

Supplier: R & D Systems
FAB7614G
102883-746EA 420.83 USD
102883-746
Anti-KLRB1C Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 694370]
Antibodies
Type: Primary
Antigen: KLRB1C
Clonality: Monoclonal
Clone: 694370
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR