Anti-CMKLR1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 477806]

Supplier: R & D Systems
FAB7610G
102883-740EA 543.75 USD
102883-740
Anti-CMKLR1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 477806]
Antibodies
Type: Primary
Antigen: CMKLR1
Clonality: Monoclonal
Clone: 477806
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR