Anti-BTLA Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 753131]

Supplier: R & D Systems
FAB7600G
102883-738EA 501.82 USD
102883-738
Anti-BTLA Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 753131]
Antibodies
Type: Primary
Antigen: BTLA
Clonality: Monoclonal
Clone: 753131
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR