Anti-TYRO3 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 109646]

Supplier: R & D Systems
FAB759G
102883-732EA 543.75 USD
102883-732
Anti-TYRO3 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 109646]
Antibodies
Type: Primary
Antigen: TYRO3
Clonality: Monoclonal
Clone: 109646
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR