Anti-S1PR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 776808]

Supplier: R & D Systems
FAB7524P
102883-720EA 513.38 USD
102883-720
Anti-S1PR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 776808]
Antibodies
Type: Primary
Antigen: S1PR3
Clonality: Monoclonal
Clone: 776808
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR