Anti-S1PR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 776808]

Supplier: R & D Systems
FAB7524A
102883-718EA 608.83 USD
102883-718
Anti-S1PR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 776808]
Antibodies
Type: Primary
Antigen: S1PR3
Clonality: Monoclonal
Clone: 776808
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR