Anti-LRIG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 789211]

Supplier: R & D Systems
FAB7498A
102883-716EA 561.11 USD
102883-716
Anti-LRIG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 789211]
Antibodies
Type: Primary
Antigen: LRIG1
Clonality: Monoclonal
Clone: 789211
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR