Anti-CD74 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 829706]

Supplier: R & D Systems
FAB7478A
102883-710EA 527.85 USD
102883-710
Anti-CD74 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 829706]
Antibodies
Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: 829706
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR