Anti-KLRB1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 702228]

Supplier: R & D Systems
FAB7448G
102883-700EA 446.86 USD
102883-700
Anti-KLRB1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 702228]
Antibodies
Type: Primary
Antigen: KLRB1
Clonality: Monoclonal
Clone: 702228
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR