Anti-FLT4 Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB743G
102883-688EA 561.11 USD
102883-688
Anti-FLT4 Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen Symbol: FLT4
Antigen Name: FMS-like tyrosine kinase 4
Antigen Synonyms: AI323512, Chy, Flt-4, VEGFR-3, VEGFR3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR