Anti-SIRPA Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 772734]

Supplier: R & D Systems
FAB7307G
102883-668EA 561.11 USD
102883-668
Anti-SIRPA Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 772734]
Antibodies
Type: Primary
Antigen: SIRPA
Clonality: Monoclonal
Clone: 772734
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR