Anti-CELSR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 763103]

Supplier: R & D Systems
FAB7278P
102883-628EA 513.38 USD
102883-628
Anti-CELSR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 763103]
Antibodies
Type: Primary
Antigen: CELSR3
Clonality: Monoclonal
Clone: 763103
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR