Anti-TIGIT Sheep Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB7267G
102883-626EA 561.11 USD
102883-626
Anti-TIGIT Sheep Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: TIGIT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR