Anti-CD34 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: QBEnd10]

Supplier: R & D Systems
FAB7227P
102883-622EA 511.94 USD
102883-622
Anti-CD34 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: QBEnd10]
Antibodies
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: QBEnd10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR