Anti-CD34 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: QBEnd10]

Supplier: R & D Systems
FAB7227G
102883-620EA 533.63 USD
102883-620
Anti-CD34 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: QBEnd10]
Antibodies
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: QBEnd10
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR