Anti-C10ORF54 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 730804]

Supplier: R & D Systems
FAB71261N
102883-596EA 577.02 USD
102883-596
Anti-C10ORF54 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 730804]
Antibodies
Type: Primary
Antigen: C10ORF54
Clonality: Monoclonal
Clone: 730804
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR