Anti-PD1 Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB7115G
102883-588EA 582.8 USD
102883-588
Anti-PD1 Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: PD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR