Anti-KLRE1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 854929]

Supplier: R & D Systems
FAB7099P
102883-586EA 513.38 USD
102883-586
Anti-KLRE1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 854929]
Antibodies
Type: Primary
Antigen: KLRE1
Clonality: Monoclonal
Clone: 854929
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR