Anti-HLAA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: W6/32]

Supplier: R & D Systems
FAB7098P
102883-582EA 582.8 USD
102883-582
Anti-HLAA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: W6/32]
Antibodies
Type: Primary
Antigen: HLAA
Clonality: Monoclonal
Clone: W6/32
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR