Anti-S1PR1 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 713412]

Supplier: R & D Systems
FAB7089N
102883-554EA 513.38 USD
102883-554
Anti-S1PR1 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 713412]
Antibodies
Type: Primary
Antigen: S1PR1
Clonality: Monoclonal
Clone: 713412
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR