Anti-CD8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: JG12]

Supplier: R & D Systems
FAB7080P
102883-548EA 323.94 USD
102883-548
Anti-CD8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: JG12]
Antibodies
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: JG12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR