Anti-CD1A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 703217]

Supplier: R & D Systems
FAB7076P
102883-542EA 420.83 USD
102883-542
Anti-CD1A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 703217]
Antibodies
Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: 703217
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR