Anti-CD1A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 703217]

Supplier: R & D Systems
FAB7076A
102883-534EA 535.08 USD
102883-534
Anti-CD1A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 703217]
Antibodies
Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: 703217
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR