Anti-JAM3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 209628]

Supplier: R & D Systems
FAB7050A
102883-530EA 594.37 USD
102883-530
Anti-JAM3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 209628]
Antibodies
Type: Primary
Antigen: JAM3
Clonality: Monoclonal
Clone: 209628
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR