Anti-VSIR Sheep Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB7005G
102883-520EA 535.08 USD
102883-520
Anti-VSIR Sheep Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: VSIR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR