Anti-ICOS Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 669222]

Supplier: R & D Systems
FAB6975A
102883-262EA 566.89 USD
102883-262
Anti-ICOS Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 669222]
Antibodies
Type: Primary
Antigen: ICOS
Clonality: Monoclonal
Clone: 669222
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR