Anti-TNFSF18 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 109101]

Supplier: R & D Systems
FAB6941N
102883-250EA 561.11 USD
102883-250
Anti-TNFSF18 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 109101]
Antibodies
Type: Primary
Antigen: TNFSF18
Clonality: Monoclonal
Clone: 109101
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR