Anti-CD79A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 706931]

Supplier: R & D Systems
FAB69201P
102883-510EA 454.09 USD
102883-510
Anti-CD79A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 706931]
Antibodies
Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: 706931
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR