Anti-CD79A Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 706931]

Supplier: R & D Systems
FAB69201G
102883-508EA 513.38 USD
102883-508
Anti-CD79A Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 706931]
Antibodies
Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: 706931
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR