Anti-CD79A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 706931]

Supplier: R & D Systems
FAB69201A
102883-506EA 561.11 USD
102883-506
Anti-CD79A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 706931]
Antibodies
Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: 706931
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR