Anti-PVR Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 690912]

Supplier: R & D Systems
FAB6909P
102883-456EA 438.18 USD
102883-456
Anti-PVR Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 690912]
Antibodies
Type: Primary
Antigen: PVR
Clonality: Monoclonal
Clone: 690912
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR