Anti-TNFRSF18 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 110416]

Supplier: R & D Systems
FAB689A-100
MSPP-FAB689A10EA 608.83 USD
MSPP-FAB689A10
Anti-TNFRSF18 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 110416]
Antibodies
Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: 110416
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR