Anti-TNFSF10 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 75402]

Supplier: R & D Systems
FAB687P
220017-963EA 513.38 USD
220017-963
Anti-TNFSF10 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 75402]
Antibodies
Type: Primary
Antigen: TNFSF10
Clonality: Monoclonal
Clone: 75402
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR