Anti-TNFSF10 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 75402]

Supplier: R & D Systems
FAB687A
102883-080EA 577.02 USD
102883-080
Anti-TNFSF10 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 75402]
Antibodies
Type: Primary
Antigen: TNFSF10
Clonality: Monoclonal
Clone: 75402
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR