Anti-KLRC1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 705829]

Supplier: R & D Systems
FAB6867G-100 FAB6867G-025
102883-432EA 561.11 USD
102883-432 102883-430
Anti-KLRC1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 705829]
Antibodies
Type: Primary
Antigen: KLRC1
Clonality: Monoclonal
Clone: 705829
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR