Anti-FGFR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 98725]

Supplier: R & D Systems
FAB684A
220017-960EA 582.8 USD
220017-960
Anti-FGFR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 98725]
Antibodies
Type: Primary
Antigen Symbol: FGFR2
Antigen Name: fibroblast growth factor receptor 2
Antigen Synonyms: BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
Clonality: Monoclonal
Clone: 98725
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR