Anti-TNFRSF11A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 80704]

Supplier: R & D Systems
FAB683P
220017-959EA 543.75 USD
220017-959
Anti-TNFRSF11A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 80704]
Antibodies
Type: Primary
Antigen: TNFRSF11A
Clonality: Monoclonal
Clone: 80704
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR