Anti-IL1RAP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 89412]

Supplier: R & D Systems
FAB676P
102883-404EA 582.8 USD
102883-404
Anti-IL1RAP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 89412]
Antibodies
Type: Primary
Antigen: IL1RAP
Clonality: Monoclonal
Clone: 89412
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR