Anti-FCRL5 Sheep Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB6757G
102883-390EA 543.75 USD
102883-390
Anti-FCRL5 Sheep Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen Symbol: FCRL5
Antigen Name: Fc receptor-like 5
Antigen Synonyms: Fcrh3, mBXMH2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR