Anti-FCRL5 Sheep Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB6757A
102883-388EA 561.11 USD
102883-388
Anti-FCRL5 Sheep Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen Symbol: FCRL5
Antigen Name: Fc receptor-like 5
Antigen Synonyms: Fcrh3, mBXMH2
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR