Anti-FCRL6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 672505]

Supplier: R & D Systems
FAB6745A
102883-374EA 594.37 USD
102883-374
Anti-FCRL6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 672505]
Antibodies
Type: Primary
Antigen Symbol: FCRL6
Antigen Name: Fc receptor-like 6
Antigen Synonyms: FcRH6
Clonality: Monoclonal
Clone: 672505
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR