Anti-IFNGR1 Mouse Monoclonal Antibody (Fluorescein) [clone: 92101]

Supplier: R & D Systems
FAB673F
220017-957EA 339.85 USD
220017-957
Anti-IFNGR1 Mouse Monoclonal Antibody (Fluorescein) [clone: 92101]
Antibodies
Type: Primary
Antigen: IFNGR1
Clonality: Monoclonal
Clone: 92101
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR