Anti-TLR5 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 624915]

Supplier: R & D Systems
FAB6704G
102883-362EA 543.75 USD
102883-362
Anti-TLR5 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 624915]
Antibodies
Type: Primary
Antigen: TLR5
Clonality: Monoclonal
Clone: 624915
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR