Anti-CD160 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 688327]

Supplier: R & D Systems
FAB6700P
102883-360EA 485.91 USD
102883-360
Anti-CD160 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 688327]
Antibodies
Type: Primary
Antigen: CD160
Clonality: Monoclonal
Clone: 688327
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR