Anti-ACKR1 Sheep Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB6695P
102883-352EA 594.37 USD
102883-352
Anti-ACKR1 Sheep Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: ACKR1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR