Anti-TACR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 694501]

Supplier: R & D Systems
FAB66871A
102883-348EA 608.83 USD
102883-348
Anti-TACR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 694501]
Antibodies
Type: Primary
Antigen: TACR1
Clonality: Monoclonal
Clone: 694501
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR