Anti-CD226 Mouse Monoclonal Antibody (Fluorescein) [clone: 102511]

Supplier: R & D Systems
FAB666F
220017-955EA 438.18 USD
220017-955
Anti-CD226 Mouse Monoclonal Antibody (Fluorescein) [clone: 102511]
Antibodies
Type: Primary
Antigen: CD226
Clonality: Monoclonal
Clone: 102511
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR