Anti-CD226 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 102511]

Supplier: R & D Systems
FAB666A
220017-954EA 594.37 USD
220017-954
Anti-CD226 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 102511]
Antibodies
Type: Primary
Antigen: CD226
Clonality: Monoclonal
Clone: 102511
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR