Anti-TNFSF14 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 115520]

Supplier: R & D Systems
FAB664A
220017-952EA 594.37 USD
220017-952
Anti-TNFSF14 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 115520]
Antibodies
Type: Primary
Antigen: TNFSF14
Clonality: Monoclonal
Clone: 115520
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR