Anti-IL1R2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 34141]

Supplier: R & D Systems
FAB663N
102883-324EA 543.75 USD
102883-324
Anti-IL1R2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 34141]
Antibodies
Type: Primary
Antigen: IL1R2
Clonality: Monoclonal
Clone: 34141
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR